Methyl-2-ethoxybenzoate 95+%

BLDPBD138272-1GEA 9.4 EUR
BLDPBD138272-1G BLDPBD138272-5G BLDPBD138272-25G BLDPBD138272-100G
Methyl-2-ethoxybenzoate 95+%
Methyl-2-ethoxybenzoate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₂O₃
MW: 180,2005 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00017261
CAS-numero: 3686-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR