Dansyylikloridi 97%

BLDPBD13851-5GEA 85.2 EUR
BLDPBD13851-5G BLDPBD13851-10G BLDPBD13851-1G BLDPBD13851-25G BLDPBD13851-250MG
Dansyylikloridi 97%
Dansyylikloridi

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₁₂H₁₂ClNO₂S
MW: 269,75 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00003985
CAS-numero: 605-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR