Methyl-3-amino-5-hydroxybenzoate 95%

BLDPBD138590-1GEA 67.6 EUR
BLDPBD138590-1G BLDPBD138590-250MG BLDPBD138590-5G
Methyl-3-amino-5-hydroxybenzoate 95%
Methyl-3-amino-5-hydroxybenzoate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₈H₉NO₃
MW: 167,1620 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09263220
CAS-numero: 67973-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR