N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 97%
|
|
BLDPBD141335-10GEA
71.7
EUR
BLDPBD141335-10G
BLDPBD141335-5G
BLDPBD141335-100G
BLDPBD141335-250MG
BLDPBD141335-25G
BLDPBD141335-1G
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide 97%
N-(4-Chloro-3-cyano-7-ethoxy-6-quinolinyl)acetamide
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Kaava:
C₁₄H₁₂ClN₃O₂ MW: 289,72 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD09263704 CAS-numero: 848133-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...