n-Octanoic anhydride 95%

BLDPBD143824-25GEA 43.7 EUR
BLDPBD143824-25G BLDPBD143824-5G
n-Octanoic anhydride 95%
n-Octanoic anhydride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₆H₃₀O₃
MW: 270,41 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00056161
CAS-numero: 623-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR