Nikkelibis(sulfamidaatti)tetrahydraatii 98+%

BLDPBD144664-25GEA 6.3 EUR
BLDPBD144664-25G
Nikkelibis(sulfamidaatti)tetrahydraatii 98+%
Nikkelibis(sulfamidaatti)tetrahydraatii

Storage condition: Inert atmosphere, Room Temperature

Kaava: H₄N₂NiO₆S₂·4H₂O
MW: 322,92 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00149966
CAS-numero: 124594-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR