9-Methylcarbazole 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD14573-1GEA 19.8 EUR
BLDPBD14573-1G BLDPBD14573-5G BLDPBD14573-25G
9-Methylcarbazole 97%
9-Methylcarbazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₃H₁₁N
MW: 181,2331 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00013431
CAS-numero: 1484-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR