o-Tolylacetonitrile 98%

BLDPBD146368-500GEA 91.5 EUR
BLDPBD146368-500G BLDPBD146368-25G BLDPBD146368-100G BLDPBD146368-10G BLDPBD146368-1000G
o-Tolylacetonitrile 98%
o-Tolylacetonitrile

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₉N
MW: 131,18 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00001904
CAS-numero: 22364-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR