Potassium-4-carboxyphenyltrifluoroborate 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD148674-5GEA 42.6 EUR
BLDPBD148674-5G BLDPBD148674-1G BLDPBD148674-25G
Potassium-4-carboxyphenyltrifluoroborate 97%
Potassium-4-carboxyphenyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₅BF₃KO₂
MW: 228,02 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04115737
CAS-numero: 850623-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR