Potassium-4-formylphenyltrifluoroborate 98%

BLDPBD148721-100GEA 517 EUR
BLDPBD148721-100G BLDPBD148721-1G BLDPBD148721-5G BLDPBD148721-25G
Potassium-4-formylphenyltrifluoroborate 98%
Potassium-4-formylphenyltrifluoroborate

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₇H₅BF₃KO
MW: 212,02 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02093980
CAS-numero: 374564-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR