4-(Diethylamino)azobenzene 98%

BLDPBD151732-500GEA 133 EUR
BLDPBD151732-0.025 BLDPBD151732-500G BLDPBD151732-1000G BLDPBD151732-25G BLDPBD151732-100G
4-(Diethylamino)azobenzene 98%
4-(Diethylamino)azobenzene

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₆H₁₉N₃
MW: 253,35 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00009043
CAS-numero: 2481-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR