4-(Bromomethyl)benzophenone 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD15192-25GEA 312 EUR
BLDPBD15192-25G BLDPBD15192-1G BLDPBD15192-5G BLDPBD15192-10G
4-(Bromomethyl)benzophenone 97%
4-(Bromomethyl)benzophenone

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₄H₁₁BrO
MW: 275,1405 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00092076
CAS-numero: 32752-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR