2,4,6-Triphenylpyridine 98%

BLDPBD1534-25GEA 1020 EUR
BLDPBD1534-25G BLDPBD1534-0.025G BLDPBD1534-1G BLDPBD1534-250MG BLDPBD1534-5G
2,4,6-Triphenylpyridine 98%
2,4,6-Triphenylpyridine

Storage condition: Sealed in dry, Room Temperature

Kaava: C₂₃H₁₇N
MW: 307,40 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00014630
CAS-numero: 580-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR