Tetrapropyyliammoniumjodidi 98%

BLDPBD153703-5GEA 9.4 EUR
BLDPBD153703-5G BLDPBD153703-25G
Tetrapropyyliammoniumjodidi 98%
Tetrapropyyliammoniumjodidi

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₂H₂₈IN
MW: 313,27 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011842
CAS-numero: 631-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR