trans,trans-4-Pentyl-4'-vinylbicyclohexyl 98%

BLDPBD154526-5GEA 36.4 EUR
BLDPBD154526-5G BLDPBD154526-1G BLDPBD154526-10G BLDPBD154526-25G
trans,trans-4-Pentyl-4'-vinylbicyclohexyl 98%
trans,trans-4-Pentyl-4'-vinylbicyclohexyl

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₉H₃₄
MW: 262,48 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11053467
CAS-numero: 129738-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR