6-Fluoroquinoline 97%
|
|
BLDPBD157697-25GEA
163
EUR
BLDPBD157697-25G
BLDPBD157697-10G
BLDPBD157697-5G
BLDPBD157697-1G
BLDPBD157697-100G
6-Fluoroquinoline 97%
6-Fluoroquinoline
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Kaava:
C₉H₆FN MW: 147,15 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD01685512 CAS-numero: 396-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...