1,2,4-Triazole-3-carboxylic acid 95%
|
|
BLDPBD157874-250MGEA
9.4
EUR
BLDPBD157874-250MG
BLDPBD157874-100G
BLDPBD157874-1G
BLDPBD157874-5G
BLDPBD157874-25G
1,2,4-Triazole-3-carboxylic acid 95%
1,2,4-Triazole-3-carboxylic acid
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₃H₃N₃O₂ MW: 113,0748 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00078268 CAS-numero: 4928-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...