3-Bromo-1H-pyrazolo[4,3-c]pyridine 97%

BLDPBD158458-250MGEA 55.1 EUR
BLDPBD158458-250MG BLDPBD158458-1G BLDPBD158458-10G BLDPBD158458-100MG BLDPBD158458-5G
3-Bromo-1H-pyrazolo[4,3-c]pyridine 97%
3-Bromo-1H-pyrazolo[4,3-c]pyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₆H₄BrN₃
MW: 198,0200 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD11518973
CAS-numero: 633328-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR