4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid 98%

BLDPBD158596-250MGEA 33.3 EUR
BLDPBD158596-250MG BLDPBD158596-1G BLDPBD158596-5G
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid 98%
4-Chlor-3-(isopropylcarbamoyl)phenylboronic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₀H₁₃BClNO₃
MW: 241,48 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07783866
CAS-numero: 871332-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR