7-Bromo-1-methyl-1H-indazole 97%

BLDPBD158846-250MGEA 35.4 EUR
BLDPBD158846-250MG BLDPBD158846-1G BLDPBD158846-25G BLDPBD158846-5G
7-Bromo-1-methyl-1H-indazole 97%
7-Bromo-1-methyl-1H-indazole

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09878570
CAS-numero: 1000576-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR