3-Amino-2-bromo-5-methoxypyridine 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD159330-5GEA 484 EUR
BLDPBD159330-5G BLDPBD159330-100MG BLDPBD159330-1G BLDPBD159330-250MG
3-Amino-2-bromo-5-methoxypyridine 98%
3-Amino-2-bromo-5-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₇BrN₂O
MW: 203,0366 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD16659663
CAS-numero: 1043688-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR