5-Bromoindan-2-ol 97%

BLDPBD160266-250MGEA 40.6 EUR
BLDPBD160266-250MG BLDPBD160266-100MG BLDPBD160266-1G
5-Bromoindan-2-ol 97%
5-Bromoindan-2-ol
Kaava: C₉H₉BrO
MW: 213,0700 g/mol
MDL numero: MFCD12755975
CAS-numero: 862135-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR