3-Fluoro-5-nitrobenzaldehyde 97%
|
|
BLDPBD160619-1GEA
41.6
EUR
BLDPBD160619-1G
BLDPBD160619-25G
BLDPBD160619-10G
BLDPBD160619-250MG
BLDPBD160619-5G
3-Fluoro-5-nitrobenzaldehyde 97%
3-Fluoro-5-nitrobenzaldehyde
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₇H₄FNO₃ MW: 169,1100 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD11111000 CAS-numero: 108159-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...