5-Bromo-2,4-difluorotoluene 98%
Toimittaja: BLD PHARMATECH GMBH
|
|
BLDPBD160871-1GEA
7.3
EUR
BLDPBD160871-1G
BLDPBD160871-5G
5-Bromo-2,4-difluorotoluene 98%
5-Bromo-2,4-difluorotoluene
Storage condition: Sealed in dry, Room Temperature
Kaava:
C₇H₅BrF₂ MW: 207,02 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD15144620 CAS-numero: 159277-47-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...