2,3-Dimethyl-6-nitro-1H-indazole 97%

BLDPBD161906-100GEA 147 EUR
BLDPBD161906-100G BLDPBD161906-25G BLDPBD161906-5G BLDPBD161906-10G
2,3-Dimethyl-6-nitro-1H-indazole 97%
2,3-Dimethyl-6-nitro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₉H₁₁N₃O₂
MW: 191,1867 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD10703326
CAS-numero: 444731-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR