1-Methyl-3-nitro-1H-pyrazole 98%

BLDPBD162200-5GEA 238 EUR
BLDPBD162200-5G BLDPBD162200-1G BLDPBD162200-250MG BLDPBD162200-25G
1-Methyl-3-nitro-1H-pyrazole 98%
1-Methyl-3-nitro-1H-pyrazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₄H₅N₃O₂
MW: 127,101 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03444368
CAS-numero: 54210-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR