2,5-Dibromoterephthalaldehyde 97%

BLDPBD164199-1GEA 67.6 EUR
BLDPBD164199-1G BLDPBD164199-250MG BLDPBD164199-100MG BLDPBD164199-10G BLDPBD164199-25G BLDPBD164199-5G
2,5-Dibromoterephthalaldehyde 97%
2,5-Dibromoterephthalaldehyde

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₄Br₂O₂
MW: 291,93 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD11521314
CAS-numero: 63525-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR