2-(Trifluoromethyl)pyridin-4-ol 97%

BLDPBD164622-10GEA 91.5 EUR
BLDPBD164622-10G BLDPBD164622-1G BLDPBD164622-100G BLDPBD164622-25G BLDPBD164622-250MG BLDPBD164622-5G
2-(Trifluoromethyl)pyridin-4-ol 97%
2-(Trifluoromethyl)pyridin-4-ol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₆H₄F₃NO
MW: 163,10 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01571202
CAS-numero: 170886-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR