7-Bromoimidazo[1,2-a]pyridine 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD164786-25GEA 472 EUR
BLDPBD164786-25G BLDPBD164786-250MG BLDPBD164786-100G BLDPBD164786-5G BLDPBD164786-1G
7-Bromoimidazo[1,2-a]pyridine 95%
7-Bromoimidazo[1,2-a]pyridine

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅BrN₂
MW: 197,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07778353
CAS-numero: 808744-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR