2-Isopropoxy-5-pyridineboronic acid 97%

BLDPBD165087-1GEA 46.8 EUR
BLDPBD165087-1G BLDPBD165087-5G BLDPBD165087-250MG
2-Isopropoxy-5-pyridineboronic acid 97%
2-Isopropoxy-5-pyridineboronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₁₂BNO₃
MW: 181,00 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07368870
CAS-numero: 870521-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR