(R)-1,1'-Spirobiindane-7,7'-diol 98% 99%ee

BLDPBD166007-250MGEA 41.6 EUR
BLDPBD166007-250MG BLDPBD166007-5G BLDPBD166007-100MG BLDPBD166007-25G BLDPBD166007-1G
(R)-1,1'-Spirobiindane-7,7'-diol 98% 99%ee
(R)-1,1'-Spirobiindane-7,7'-diol

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₇H₁₆O₂
MW: 252,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD31714175
CAS-numero: 223259-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR