1-Methyl-1H-indazole-4-boronic acid 98%

BLDPBD167731-250MGEA 43.7 EUR
BLDPBD167731-250MG BLDPBD167731-5G BLDPBD167731-1G
1-Methyl-1H-indazole-4-boronic acid 98%
1-Methyl-1H-indazole-4-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₈H₉BN₂O₂
MW: 175,9803 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09870045
CAS-numero: 1001907-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR