(2-Bromo-5-fluorophenyl)acetic acid 98%

BLDPBD168810-250MGEA 7.3 EUR
BLDPBD168810-250MG BLDPBD168810-5G BLDPBD168810-100G BLDPBD168810-1G BLDPBD168810-25G BLDPBD168810-10G
(2-Bromo-5-fluorophenyl)acetic acid 98%
(2-Bromo-5-fluorophenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₆BrFO₂
MW: 233,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03094440
CAS-numero: 739336-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR