MOPS 97%

BLDPBD17201-1000GEA 99.8 EUR
BLDPBD17201-1000G BLDPBD17201-100G BLDPBD17201-500G BLDPBD17201-25G
MOPS 97%
MOPS

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₇H₁₅NO₄S
MW: 209,27 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006183
CAS-numero: 1132-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR