3-Bromo-4-hydroxy-5-methoxybenzonitrile 98%
|
|
BLDPBD172344-5GEA
192
EUR
BLDPBD172344-5G
BLDPBD172344-1G
BLDPBD172344-250MG
3-Bromo-4-hydroxy-5-methoxybenzonitrile 98%
3-Bromo-4-hydroxy-5-methoxybenzonitrile
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₈H₆BrNO₂ MW: 228,0400 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD01123027 CAS-numero: 52805-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...