3-Bromo-4-(methylsulphonyl)benzoic acid 97%

BLDPBD174183-250MGEA 44.7 EUR
BLDPBD174183-250MG BLDPBD174183-5G BLDPBD174183-100MG BLDPBD174183-1G
3-Bromo-4-(methylsulphonyl)benzoic acid 97%
3-Bromo-4-(methylsulphonyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₇BrO₄S
MW: 279,1100 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD13191607
CAS-numero: 39058-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR