(±)-1-Benzyl-3-hydroxypiperidine hydrochloride 98+%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD17430-5GEA 26 EUR
BLDPBD17430-5G BLDPBD17430-1G BLDPBD17430-25G BLDPBD17430-100G
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride 98+%
(±)-1-Benzyl-3-hydroxypiperidine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₂H₁₈ClNO
MW: 227,7304 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00044200
CAS-numero: 105973-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR