N-Methoxycarbonyl-D-phenylglycine 97%

BLDPBD17638-100GEA 31.2 EUR
BLDPBD17638-100G BLDPBD17638-10G BLDPBD17638-25G
N-Methoxycarbonyl-D-phenylglycine 97%
N-Methoxycarbonyl-D-phenylglycine

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₁₁NO₄
MW: 209,20 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD12796004
CAS-numero: 50890-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR