(R)-3-(Boc-amino)-3-phenylpropionic acid 97%

BLDPBD18679-5GEA 154 EUR
BLDPBD18679-5G BLDPBD18679-25G BLDPBD18679-100MG BLDPBD18679-250MG BLDPBD18679-1G
(R)-3-(Boc-amino)-3-phenylpropionic acid 97%
(R)-3-(Boc-amino)-3-phenylpropionic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₄H₁₉NO₄
MW: 265,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD01320859
CAS-numero: 161024-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR