4-(Diethylamino)salicylaldehyde 98%

BLDPBD18922-100GEA 22.9 EUR
BLDPBD18922-100G BLDPBD18922-1000G BLDPBD18922-500G BLDPBD18922-25G
4-(Diethylamino)salicylaldehyde 98%
4-(Diethylamino)salicylaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₁H₁₅NO₂
MW: 193,25 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00003326
CAS-numero: 17754-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR