4,5,6-Trichloropyrimidine 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD18933-1GEA 44.7 EUR
BLDPBD18933-1G BLDPBD18933-100MG BLDPBD18933-250MG BLDPBD18933-5G BLDPBD18933-25G BLDPBD18933-100G
4,5,6-Trichloropyrimidine 97%
4,5,6-Trichloropyrimidine

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₄HCl₃N₂
MW: 183,4231 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08458457
CAS-numero: 1780-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR