tert-Butyl 2-bromoisobutyrate 97%
|
|
BLDPBD19576-100GEA
27.1
EUR
BLDPBD19576-100G
BLDPBD19576-500G
BLDPBD19576-1000G
BLDPBD19576-25G
BLDPBD19576-5G
tert-Butyl 2-bromoisobutyrate 97%
tert-Butyl 2-bromoisobutyrate
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₆H₁₁BrO₂ MW: 223,12 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00051908 CAS-numero: 23877-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...