Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD196680-250MGEA 22.9 EUR
BLDPBD196680-250MG BLDPBD196680-5G BLDPBD196680-1G
Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate 98%
Ethyl-2,7-dimethylimidazo[1,2-a]pyridine-3-carboxylate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₂H₁₄N₂O₂
MW: 218,2500 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00139510
CAS-numero: 81448-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR