4-Fluoro-7-nitrobenzofurazan 95%

BLDPBD20023-100MGEA 144 EUR
BLDPBD20023-100MG BLDPBD20023-1G BLDPBD20023-250MG
4-Fluoro-7-nitrobenzofurazan 95%
4-Fluoro-7-nitrobenzofurazan

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₆H₂FN₃O₃
MW: 183,1 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00010196
CAS-numero: 29270-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR