4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD201103-5GEA 383 EUR
BLDPBD201103-5G BLDPBD201103-250MG BLDPBD201103-1G BLDPBD201103-100MG
4-Bromo-2-fluoro-5-nitrobenzaldehyde 95%
4-Bromo-2-fluoro-5-nitrobenzaldehyde

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₇H₃BrFNO₃
MW: 248,0060 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD10758067
CAS-numero: 679839-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR