4-Chloro-2-picoline 98%

BLDPBD20479-5GEA 17.7 EUR
BLDPBD20479-5G BLDPBD20479-10G BLDPBD20479-1G BLDPBD20479-25G BLDPBD20479-100G
4-Chloro-2-picoline 98%
4-Chloro-2-picoline

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₆H₆ClN
MW: 127,57 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00038833
CAS-numero: 3678-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR