5-Amino-2,4,6-triiodoisophthaloyl Dichloride 98%

BLDPBD20514-5GEA 55.1 EUR
BLDPBD20514-5G BLDPBD20514-1G BLDPBD20514-25G
5-Amino-2,4,6-triiodoisophthaloyl Dichloride 98%
5-Amino-2,4,6-triiodoisophthaloyl Dichloride

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₈H₂Cl₂I₃NO₂
MW: 595,72 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00270836
CAS-numero: 37441-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR