4-Nitro-2,3-lutidine-N-oxide 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD20541-5GEA 7.3 EUR
BLDPBD20541-5G BLDPBD20541-100G BLDPBD20541-25G BLDPBD20541-500G
4-Nitro-2,3-lutidine-N-oxide 97%
4-Nitro-2,3-lutidine-N-oxide

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₈N₂O₂
MW: 168,15 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00065172
CAS-numero: 37699-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR