4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD208200-100MGEA 25 EUR
BLDPBD208200-100MG BLDPBD208200-1G BLDPBD208200-250MG BLDPBD208200-5G
4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride 95%
4,5-Dichloro-1,2,3-dithiazol-2-ylium chloride

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₂Cl₂NS₂·Cl
MW: 208,517 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD03426194
CAS-numero: 75318-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR