2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%

BLDPBD208567-250MGEA 38.5 EUR
BLDPBD208567-250MG BLDPBD208567-5G BLDPBD208567-1G BLDPBD208567-100MG
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 95%
2-Amino-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Kaava: C₁₂H₁₆BF₃N₂O₂
MW: 288,07 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD12923414
CAS-numero: 947249-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR